Assay Details
| Gene Symbol: NF-YC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104356.3 | NP_175880.1 | 2 - 3 | 914 | 127 | |
| NM_179477.2 | NP_849808.1 | 1 - 2 | 737 | 127 | ||
| GenBank mRNA | AY062775.1 | - | 2 - 3 | 765 | 127 | |
| AY081654.1 | - | 1 - 2 | 674 | 127 | ||
| AY087517.1 | - | 2 - 3 | 766 | 127 | ||
More Information
Set Membership: |
|

NF-YC3
Genomic Map