Assay Details
| Gene Symbol: LOX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104376.3 | NP_175900.1 | 1 - 2 | 338 | 84 | |
| GenBank mRNA | AY093104.1 | - | 1 - 2 | 282 | 84 | |
| BT010358.1 | - | 1 - 2 | 194 | 84 | ||
| L04637.1 | - | 1 - 2 | 274 | 84 | ||
More Information
Set Membership: |
|

LOX1
Genomic Map