Assay Details
| Gene Symbol: LOX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104376.3 | NP_175900.1 | 3 - 4 | 1214 | 82 | |
| GenBank mRNA | AY093104.1 | - | 3 - 4 | 1158 | 82 | |
| BT010358.1 | - | 3 - 4 | 1070 | 82 | ||
| L04637.1 | - | 3 - 4 | 1150 | 82 | ||
More Information
Set Membership: |
|

LOX1
Genomic Map