Assay Details
| Gene Symbol: LOX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104376.3 | NP_175900.1 | 7 - 8 | 1976 | 65 | |
| GenBank mRNA | AY093104.1 | - | 7 - 8 | 1920 | 65 | |
| BT010358.1 | - | 7 - 8 | 1832 | 65 | ||
| L04637.1 | - | 7 - 8 | 1912 | 65 | ||
More Information
Set Membership: |
|

LOX1
Genomic Map