Assay Details
| Gene Symbol: RH20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104388.4 | NP_175911.1 | 1 - 2 | 215 | 88 | |
| GenBank mRNA | AY062745.1 | - | 1 - 2 | 211 | 88 | |
| AY128786.1 | - | 1 - 2 | 61 | 88 | ||
More Information
Set Membership: |
|

RH20
Genomic Map