Assay Details
| Gene Symbol: RH20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001333691.1 | NP_001322402.1 | 2 - 3 | 854 | 112 | |
| NM_104388.4 | NP_175911.1 | 3 - 4 | 658 | 112 | ||
| GenBank mRNA | AY062745.1 | - | 3 - 4 | 654 | 112 | |
| AY128786.1 | - | 3 - 4 | 504 | 112 | ||
More Information
Set Membership: |
|

RH20
Genomic Map