Assay Details
| Gene Symbol: RH20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001333691.1 | NP_001322402.1 | 8 - 9 | 1811 | 64 | |
| NM_104388.4 | NP_175911.1 | 9 - 10 | 1615 | 64 | ||
| GenBank mRNA | AY062745.1 | - | 9 - 10 | 1611 | 64 | |
| AY128786.1 | - | 9 - 10 | 1461 | 64 | ||
More Information
Set Membership: |
|

RH20
Genomic Map