Assay Details
| Gene Symbol: TAF7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036117.1 | NP_001031194.1 | 1 - 2 | 321 | 85 | |
| NM_104405.4 | NP_175926.1 | 1 - 2 | 321 | 85 | ||
| GenBank mRNA | AK316930.1 | - | 1 - 2 | 302 | 85 | |
| AK316996.1 | - | 1 - 2 | 283 | 85 | ||
| AY463622.1 | - | 1 - 2 | 119 | 85 | ||
| BT011002.1 | - | 1 - 2 | 246 | 85 | ||
| BT015099.1 | - | 1 - 2 | 119 | 85 | ||
More Information
Set Membership: |
|

TAF7
Genomic Map