Assay Details
| Gene Symbol: SCL33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036118.4 | NP_001031195.4 | 4 - 5 | 487 | 78 | |
| NM_001333699.1 | NP_001319238.1 | 4 - 5 | 720 | 78 | ||
| NM_001333700.1 | NP_001319239.1 | 5 - 6 | 518 | 78 | ||
| NM_001333701.1 | NP_001321067.1 | 3 - 4 | 1619 | 78 | ||
| NM_104406.6 | NP_564685.4 | 4 - 5 | 854 | 78 | ||
| GenBank mRNA | AF099940.2 | - | 4 - 5 | 457 | 78 | |
| AJ293799.1 | - | 4 - 5 | 455 | 78 | ||
| AK318976.1 | - | 5 - 6 | 632 | 78 | ||
| BT020420.1 | - | 3 - 4 | 396 | 78 | ||
More Information
Set Membership: |
|

SCL33
Genomic Map