Assay Details
| Gene Symbol: BCDH BETA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104426.4 | NP_175947.1 | 11 - 12 | 1218 | 91 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

BCDH BETA1| Gene Symbol: BCDH BETA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104426.4 | NP_175947.1 | 11 - 12 | 1218 | 91 | |
Set Membership: |
|