Assay Details
| Gene Symbol: BCDH BETA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104426.4 | NP_175947.1 | 2 - 3 | 465 | 92 | |
| GenBank mRNA | BT024741.1 | - | 2 - 3 | 262 | 92 | |
More Information
Set Membership: |
|

BCDH BETA1
Genomic Map