Assay Details
| Gene Symbol: TBP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001333716.1 | NP_001321102.1 | 4 - 5 | 502 | 85 | |
| NM_001333717.1 | NP_001321103.1 | 4 - 5 | 504 | 85 | ||
| NM_104427.4 | NP_175948.1 | 4 - 5 | 502 | 85 | ||
| NM_179481.2 | NP_849812.1 | 4 - 5 | 502 | 85 | ||
| GenBank mRNA | AB493506.1 | - | 3 - 4 | 235 | 85 | |
| AK227470.1 | - | 4 - 5 | 521 | 85 | ||
| AY463626.1 | - | 3 - 4 | 235 | 85 | ||
| BT006488.1 | - | 3 - 4 | 235 | 85 | ||
| X54995.1 | - | 489 | 85 | |||
More Information
Set Membership: |
|

TBP2
Genomic Map