Assay Details
| Gene Symbol: TBP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001333717.1 | NP_001321103.1 | 8 - 9 | 790 | 147 | |
| NM_104427.4 | NP_175948.1 | 8 - 9 | 788 | 147 | ||
| NM_179481.2 | NP_849812.1 | 8 - 9 | 788 | 147 | ||
| GenBank mRNA | AB493506.1 | - | 7 - 8 | 521 | 147 | |
| AK227470.1 | - | 8 - 9 | 807 | 147 | ||
| AY463626.1 | - | 7 - 8 | 521 | 147 | ||
| BT006488.1 | - | 7 - 8 | 521 | 147 | ||
| X54995.1 | - | 775 | 147 | |||
More Information
Set Membership: |
|

TBP2
Genomic Map