Assay Details
| Gene Symbol: SIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104450.4 | NP_175970.1 | 4 - 5 | 654 | 97 | |
| GenBank mRNA | AK227977.1 | - | 4 - 5 | 676 | 97 | |
| BT004640.1 | - | 4 - 5 | 583 | 97 | ||
More Information
Set Membership: |
|

SIP1
Genomic Map