Assay Details
| Gene Symbol: SIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104450.4 | NP_175970.1 | 7 - 8 | 1392 | 119 | |
| GenBank mRNA | AK227977.1 | - | 7 - 8 | 1414 | 119 | |
| BT004640.1 | - | 7 - 8 | 1321 | 119 | ||
More Information
Set Membership: |
|

SIP1
Genomic Map