Assay Details
| Gene Symbol: AT1G55805 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104457.5 | NP_564702.2 | 1 | 597 | 150 | |
| GenBank mRNA | AK175386.1 | - | 1 | 553 | 150 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT1G55805| Gene Symbol: AT1G55805 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104457.5 | NP_564702.2 | 1 | 597 | 150 | |
| GenBank mRNA | AK175386.1 | - | 1 | 553 | 150 | |
Set Membership: |
|