Assay Details
| Gene Symbol: AHG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001333741.1 | NP_001322687.1 | 2 - 3 | 578 | 118 | |
| NM_104464.7 | NP_175983.5 | 2 - 3 | 578 | 118 | ||
| GenBank mRNA | AB194966.1 | - | 2 - 3 | 497 | 118 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001333741.1 transcripts.
Set Membership: |
|

AHG2
Genomic Map