Assay Details
| Gene Symbol: AHG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001333740.1 | NP_001319244.1 | 4 - 5 | 1189 | 110 | |
| NM_001333741.1 | NP_001322687.1 | 4 - 5 | 1211 | 110 | ||
| NM_001333742.1 | NP_001322686.1 | 5 - 6 | 1117 | 110 | ||
| NM_104464.7 | NP_175983.5 | 5 - 6 | 1136 | 110 | ||
| GenBank mRNA | AB194966.1 | - | 5 - 6 | 1055 | 110 | |
More Information
Set Membership: |
|

AHG2
Genomic Map