Assay Details
| Gene Symbol: TIM50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104467.5 | NP_175986.2 | 9 - 10 | 1184 | 71 | |
| GenBank mRNA | AY072140.1 | - | 9 - 10 | 1181 | 71 | |
| AY122975.1 | - | 9 - 10 | 1087 | 71 | ||
More Information
Set Membership: |
|

TIM50
Genomic Map