Assay Details
| Gene Symbol: NOP56 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104489.4 | NP_176007.1 | 4 - 5 | 827 | 77 | |
| GenBank mRNA | AF302492.1 | - | 4 - 5 | 768 | 77 | |
| AY039541.1 | - | 4 - 5 | 822 | 77 | ||
| AY087080.1 | - | 4 - 5 | 824 | 77 | ||
| AY102151.1 | - | 4 - 5 | 768 | 77 | ||
More Information
Set Membership: |
|

NOP56
Genomic Map