Assay Details
| Gene Symbol: CRT1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036122.2 | NP_001031199.1 | 3 - 4 | 424 | 89 | |
| NM_104513.5 | NP_176030.1 | 3 - 4 | 474 | 89 | ||
| GenBank mRNA | AY062628.1 | - | 3 - 4 | 422 | 89 | |
| BT008511.1 | - | 3 - 4 | 395 | 89 | ||
| U66343.1 | - | 3 - 4 | 398 | 89 | ||
More Information
Set Membership: |
|

CRT1a
Genomic Map