Assay Details
| Gene Symbol: CYP96A15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001124037.1 | NP_001117509.1 | 2 | 1559 | 124 | |
| NM_104570.3 | NP_176086.1 | 1 | 1645 | 124 | ||
More Information
Set Membership: |
|

CYP96A15
Genomic Map