Assay Details
| Gene Symbol: VIM5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001333805.1 | NP_001321213.1 | 1 - 2 | 512 | 66 | |
| NM_104575.3 | NP_176091.2 | 1 - 2 | 512 | 66 | ||
More Information
Set Membership: |
|

VIM5
Genomic Map