Assay Details
| Gene Symbol: VIM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001333806.1 | NP_001319260.1 | 3 - 4 | 1342 | 150 | |
| NM_104576.6 | NP_176092.2 | 3 - 4 | 1342 | 150 | ||
| NM_202316.2 | NP_974045.1 | 3 - 4 | 1155 | 150 | ||
| GenBank mRNA | AY065438.1 | - | 3 - 4 | 1155 | 150 | |
| AY117235.1 | - | 3 - 4 | 1129 | 150 | ||
More Information
Set Membership: |
|

VIM1
Genomic Map