Assay Details
| Gene Symbol: AT1G58060 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104588.4 | NP_176103.2 | 27 - 28 | 3751 | 66 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT1G58060| Gene Symbol: AT1G58060 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104588.4 | NP_176103.2 | 27 - 28 | 3751 | 66 | |
Set Membership: |
|