Assay Details
| Gene Symbol: SAB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198333.1 | NP_001185262.1 | 16 - 17 | 6785 | 73 | |
| NM_104605.4 | NP_176121.3 | 15 - 16 | 6760 | 73 | ||
More Information
Set Membership: |
|

SAB
Genomic Map