Assay Details
| Gene Symbol: SAB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198333.1 | NP_001185262.1 | 22 - 23 | 7745 | 116 | |
| NM_104605.4 | NP_176121.3 | 21 - 22 | 7720 | 116 | ||
More Information
Set Membership: |
|

SAB
Genomic Map