Assay Details
| Gene Symbol: ZW9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104607.3 | NP_564729.1 | 1 - 2 | 468 | 70 | |
| GenBank mRNA | AB028194.1 | - | 1 - 2 | 397 | 70 | |
| AF332419.1 | - | 1 - 2 | 388 | 70 | ||
| AF419580.1 | - | 1 - 2 | 414 | 70 | ||
More Information
Set Membership: |
|

ZW9
Genomic Map