Assay Details
| Gene Symbol: ZW9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104607.3 | NP_564729.1 | 4 - 5 | 1081 | 67 | |
| GenBank mRNA | AB028194.1 | - | 4 - 5 | 1010 | 67 | |
| AF332419.1 | - | 4 - 5 | 1001 | 67 | ||
| AF419580.1 | - | 4 - 5 | 1027 | 67 | ||
More Information
Set Membership: |
|

ZW9
Genomic Map