Assay Details
| Gene Symbol: ZF14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104614.5 | NP_564731.1 | 1 - 2 | 333 | 133 | |
| GenBank mRNA | AB028198.1 | - | 1 - 2 | 297 | 133 | |
More Information
Set Membership: |
|

ZF14
Genomic Map