Assay Details
| Gene Symbol: ZW18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001124042.2 | NP_001117514.1 | 13 - 14 | 2881 | 94 | |
| NM_001198335.1 | NP_001185264.1 | 13 - 14 | 2269 | 94 | ||
| NM_001333828.1 | NP_001323158.1 | 14 - 15 | 2425 | 94 | ||
| NM_104615.4 | NP_564732.1 | 14 - 15 | 2709 | 94 | ||
| GenBank mRNA | AB028199.1 | - | 14 - 15 | 2428 | 94 | |
| AK226788.1 | - | 14 - 15 | 2525 | 94 | ||
More Information
Set Membership: |
|

ZW18
Genomic Map