Assay Details
| Gene Symbol: ZW18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001124042.2 | NP_001117514.1 | 2 - 3 | 1060 | 125 | |
| NM_001198335.1 | NP_001185264.1 | 3 - 4 | 697 | 125 | ||
| NM_001333828.1 | NP_001323158.1 | 3 - 4 | 604 | 125 | ||
| NM_104615.4 | NP_564732.1 | 3 - 4 | 888 | 125 | ||
| GenBank mRNA | AB028199.1 | - | 3 - 4 | 607 | 125 | |
More Information
Set Membership: |
|

ZW18
Genomic Map