Assay Details
| Gene Symbol: AAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104616.5 | NP_176132.1 | 2 - 3 | 674 | 105 | |
| GenBank mRNA | L16240.1 | - | 2 - 3 | 495 | 105 | |
| X67124.1 | - | 495 | 105 | |||
More Information
Set Membership: |
|

AAP1
Genomic Map