Assay Details
| Gene Symbol: AAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104616.5 | NP_176132.1 | 3 - 4 | 900 | 95 | |
| GenBank mRNA | L16240.1 | - | 3 - 4 | 721 | 95 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AAP1| Gene Symbol: AAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104616.5 | NP_176132.1 | 3 - 4 | 900 | 95 | |
| GenBank mRNA | L16240.1 | - | 3 - 4 | 721 | 95 | |
Set Membership: |
|