Assay Details
| Gene Symbol: RBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001333838.1 | NP_001320402.1 | 1 - 2 | 614 | 121 | |
| NM_104627.2 | NP_176143.1 | 2 - 3 | 365 | 121 | ||
| GenBank mRNA | AY054536.1 | - | 2 - 3 | 304 | 121 | |
| AY064643.1 | - | 1 - 2 | 221 | 121 | ||
More Information
Set Membership: |
|

RBP1
Genomic Map