Assay Details
| Gene Symbol: DL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104654.5 | NP_176170.1 | 16 - 17 | 2153 | 116 | |
| GenBank mRNA | AB028467.1 | - | 16 - 17 | 2055 | 116 | |
| AY049230.1 | - | 16 - 17 | 2071 | 116 | ||
| BT000772.1 | - | 16 - 17 | 2076 | 116 | ||
| BT009695.1 | - | 16 - 17 | 1882 | 116 | ||
More Information
Set Membership: |
|

DL3
Genomic Map