Assay Details
| Gene Symbol: DL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104654.5 | NP_176170.1 | 21 - 22 | 2797 | 65 | |
| GenBank mRNA | AB028467.1 | - | 21 - 22 | 2699 | 65 | |
| AY049230.1 | - | 21 - 22 | 2715 | 65 | ||
| BT000772.1 | - | 21 - 22 | 2720 | 65 | ||
| BT009695.1 | - | 21 - 22 | 2526 | 65 | ||
More Information
Set Membership: |
|

DL3
Genomic Map