Assay Details
| Gene Symbol: AT1G59740 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104667.3 | NP_176183.1 | 2 - 3 | 613 | 98 | |
| GenBank mRNA | AY035037.1 | - | 2 - 3 | 528 | 98 | |
| AY051041.1 | - | 2 - 3 | 372 | 98 | ||
More Information
Set Membership: |
|

AT1G59740
Genomic Map