Assay Details
| Gene Symbol: AT1G59740 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104667.3 | NP_176183.1 | 4 - 5 | 1183 | 64 | |
| GenBank mRNA | AY035037.1 | - | 4 - 5 | 1098 | 64 | |
| AY051041.1 | - | 4 - 5 | 942 | 64 | ||
More Information
Set Membership: |
|

AT1G59740
Genomic Map