Assay Details
| Gene Symbol: ARF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036131.3 | NP_001031208.1 | 8 - 9 | 750 | 63 | |
| NM_001198337.1 | NP_001185266.1 | 8 - 9 | 996 | 63 | ||
| NM_104668.4 | NP_176184.1 | 8 - 9 | 946 | 63 | ||
| NM_179499.2 | NP_849830.1 | 8 - 9 | 901 | 63 | ||
| GenBank mRNA | AK317214.1 | - | 8 - 9 | 706 | 63 | |
| BT002748.1 | - | 7 - 8 | 635 | 63 | ||
| U83245.1 | - | 8 - 9 | 749 | 63 | ||
More Information
Set Membership: |
|

ARF1
Genomic Map