Assay Details
| Gene Symbol: AT1G59850 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104678.2 | NP_176194.1 | 1 - 2 | 884 | 137 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT1G59850| Gene Symbol: AT1G59850 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104678.2 | NP_176194.1 | 1 - 2 | 884 | 137 | |
Set Membership: |
|