Assay Details
| Gene Symbol: PEN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104680.3 | NP_176196.1 | 3 - 4 | 2466 | 116 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PEN3| Gene Symbol: PEN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104680.3 | NP_176196.1 | 3 - 4 | 2466 | 116 | |
Set Membership: |
|