Assay Details
| Gene Symbol: emb1220 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104707.4 | NP_564754.1 | 6 - 7 | 1434 | 77 | |
| GenBank mRNA | AY080777.1 | - | 6 - 7 | 1419 | 77 | |
| AY084782.1 | - | 6 - 7 | 1369 | 77 | ||
| AY117155.1 | - | 4 - 5 | 1281 | 77 | ||
More Information
Set Membership: |
|

emb1220
Genomic Map