Assay Details
| Gene Symbol: PANK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001333894.1 | NP_001320517.1 | 7 - 8 | 1101 | 80 | |
| NM_001333895.1 | NP_001320518.1 | 7 - 8 | 865 | 80 | ||
| NM_104731.5 | NP_176247.2 | 8 - 9 | 997 | 80 | ||
| GenBank mRNA | AK229327.1 | - | 8 - 9 | 980 | 80 | |
| BT011005.1 | - | 8 - 9 | 847 | 80 | ||
| BT015384.1 | - | 8 - 9 | 809 | 80 | ||
More Information
Set Membership: |
|

PANK1
Genomic Map