Assay Details
| Gene Symbol: ECHID | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104739.5 | NP_176255.2 | 1 - 2 | 457 | 136 | |
| GenBank mRNA | BT033085.1 | - | 1 - 2 | 260 | 136 | |
More Information
Set Membership: |
|

ECHID
Genomic Map