Assay Details
| Gene Symbol: ECHID | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104739.5 | NP_176255.2 | 5 - 6 | 831 | 103 | |
| GenBank mRNA | BT033085.1 | - | 5 - 6 | 634 | 103 | |
More Information
Set Membership: |
|

ECHID
Genomic Map