Assay Details
| Gene Symbol: ECHID | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104739.5 | NP_176255.2 | 7 - 8 | 972 | 97 | |
| GenBank mRNA | BT033085.1 | - | 7 - 8 | 775 | 97 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ECHID| Gene Symbol: ECHID | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104739.5 | NP_176255.2 | 7 - 8 | 972 | 97 | |
| GenBank mRNA | BT033085.1 | - | 7 - 8 | 775 | 97 | |
Set Membership: |
|