Assay Details
| Gene Symbol: AGD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001333920.1 | NP_001322222.1 | 14 - 15 | 1743 | 65 | |
| NM_001333921.1 | NP_001322221.1 | 8 - 9 | 1418 | 65 | ||
| NM_104767.4 | NP_176283.1 | 16 - 17 | 1865 | 65 | ||
More Information
Set Membership: |
|

AGD2
Genomic Map