Assay Details
| Gene Symbol: AGD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001333920.1 | NP_001322222.1 | 2 - 3 | 366 | 128 | |
| NM_104767.4 | NP_176283.1 | 4 - 5 | 488 | 128 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001333920.1 transcripts.
Set Membership: |
|

AGD2
Genomic Map