Assay Details
| Gene Symbol: AGD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001333920.1 | NP_001322222.1 | 3 - 4 | 425 | 109 | |
| NM_104767.4 | NP_176283.1 | 5 - 6 | 547 | 109 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001333920.1 transcripts.
Set Membership: |
|

AGD2
Genomic Map